Product Details

SNP ID
rs143591340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66988494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCAATTTTCCACAGCTTCATCAA[C/T]TTGTTCAGTAAAAACTCCAAATCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R42 PubMed Links

Gene Details

Gene
PPP1R42
Gene Name
protein phosphatase 1 regulatory subunit 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013626.2 721 Silent Mutation AAA,AAG K192K NP_001013648.1

View Full Product Details