Product Details

SNP ID
rs143679528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33593629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGAGTCGTGGGCCTCAACACCCA[A/G]GTAGCGGATGCCCAGCCCCTCATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP26 PubMed Links

Gene Details

Gene
DUSP26
Gene Name
dual specificity phosphatase 26 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305115.1 622 Silent Mutation CTG,TTG L114L NP_001292044.1
NM_001305116.1 622 Silent Mutation CTG,TTG L114L NP_001292045.1
NM_024025.2 622 Silent Mutation CTG,TTG L114L NP_076930.1

View Full Product Details