Product Details

SNP ID
rs143735406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11331568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCTGCCTTGAGAGCCAGGGTCT[C/T]GGTGGCTACGAGTGGTGTGGACTGT
Phenotype
MIM: 606260 MIM: 615199 MIM: 615174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929290 PubMed Links

Gene Details

Gene
LOC101929290
Gene Name
uncharacterized LOC101929290
There are no transcripts associated with this gene.

Gene
MTMR9
Gene Name
myotubularin related protein 9
There are no transcripts associated with this gene.

Gene
SLC35G5
Gene Name
solute carrier family 35 member G5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054028.1 Intron NP_473369.1
Gene
TDH
Gene Name
L-threonine dehydrogenase (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details