Product Details

SNP ID
rs143763357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42344962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTTTCCTTATAGAAAAGCAGC[A/G]TCTGTTATAGCAAAATACCCACACA
Phenotype
MIM: 174760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLB PubMed Links

Gene Details

Gene
POLB
Gene Name
DNA polymerase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002690.2 296 Silent Mutation GCA,GCG A43A NP_002681.1
XM_005273535.3 296 Silent Mutation GCA,GCG A43A XP_005273592.1
XM_005273536.3 296 Silent Mutation GCA,GCG A43A XP_005273593.1
XM_005273537.3 296 Silent Mutation GCA,GCG A43A XP_005273594.1
XM_005273538.2 296 UTR 5 XP_005273595.1
XM_005273539.2 296 UTR 5 XP_005273596.1
XM_005273540.4 296 UTR 5 XP_005273597.1
XM_006716353.2 296 UTR 5 XP_006716416.1
XM_017013583.1 296 UTR 5 XP_016869072.1
XM_017013584.1 296 UTR 5 XP_016869073.1

View Full Product Details