Product Details
- SNP ID
-
rs143803245
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:119732146 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAGACTGAGAGTGGAGCGCTTGC[C/T]GCTGGATGCATACTCATCTCCAAAG
- Phenotype
-
MIM: 604912
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TAF2
PubMed Links
Gene Details
- Gene
- TAF2
- Gene Name
- TATA-box binding protein associated factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003184.3 |
3514 |
Silent Mutation |
GCA,GCG |
A1126A |
NP_003175.1 |
XM_006716621.3 |
3514 |
Silent Mutation |
GCA,GCG |
A1178A |
XP_006716684.1 |
XM_011517259.2 |
3514 |
Silent Mutation |
GCA,GCG |
A1141A |
XP_011515561.1 |
XM_017013779.1 |
3514 |
Silent Mutation |
GCA,GCG |
A1089A |
XP_016869268.1 |
XM_017013780.1 |
3514 |
Silent Mutation |
GCA,GCG |
A1009A |
XP_016869269.1 |
XM_017013781.1 |
3514 |
Silent Mutation |
GCA,GCG |
A1009A |
XP_016869270.1 |
XM_017013782.1 |
3514 |
Silent Mutation |
GCA,GCG |
A1009A |
XP_016869271.1 |
XM_017013783.1 |
3514 |
Silent Mutation |
GCA,GCG |
A967A |
XP_016869272.1 |
XM_017013784.1 |
3514 |
Silent Mutation |
GCA,GCG |
A957A |
XP_016869273.1 |
XM_017013785.1 |
3514 |
Silent Mutation |
GCA,GCG |
A957A |
XP_016869274.1 |
XM_017013786.1 |
3514 |
Silent Mutation |
GCA,GCG |
A957A |
XP_016869275.1 |
XM_017013787.1 |
3514 |
Silent Mutation |
GCA,GCG |
A920A |
XP_016869276.1 |
XM_017013788.1 |
3514 |
Silent Mutation |
GCA,GCG |
A761A |
XP_016869277.1 |
XM_017013789.1 |
3514 |
Silent Mutation |
GCA,GCG |
A709A |
XP_016869278.1 |
XM_017013790.1 |
3514 |
Silent Mutation |
GCA,GCG |
A709A |
XP_016869279.1 |
XM_017013791.1 |
3514 |
Silent Mutation |
GCA,GCG |
A679A |
XP_016869280.1 |
View Full Product Details