Product Details
- SNP ID
-
rs144147835
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:123684652 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATGCGAATCAACGTCTCCTCATC[A/G]GTCCCCGCACCCTTCATCGACTTGT
- Phenotype
-
MIM: 602573
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA13
PubMed Links
Gene Details
- Gene
- ANXA13
- Gene Name
- annexin A13
View Full Product Details