Product Details

SNP ID
rs144426877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56113782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACACCGAGCCAAACCCTCCAGC[A/T]CCCAGCCTCTGCAGCAAGCACACCT
Phenotype
MIM: 190060
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MOS PubMed Links

Gene Details

Gene
MOS
Gene Name
v-mos Moloney murine sarcoma viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005372.1 201 Silent Mutation GGA,GGT G67G NP_005363.1

View Full Product Details