Product Details

SNP ID
rs144546839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38172012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCCTATAAGTGTCCTTCCATCT[C/T]GAAGCAGAACCAAGTGCTTTTCTGG
Phenotype
MIM: 603884 MIM: 607281
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAG4 PubMed Links

Gene Details

Gene
BAG4
Gene Name
BCL2 associated athanogene 4
There are no transcripts associated with this gene.

Gene
LSM1
Gene Name
LSM1 homolog, mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014462.2 478 Missense Mutation CAA,CGA Q23R NP_055277.1

View Full Product Details