Product Details

SNP ID
rs144993866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142912003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTAGTTAATCGCTCTGAAAGTGA[G/T]GAGGGGGGACGTGCCAGGCCTCAAT
Phenotype
MIM: 124080
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP11B2 PubMed Links

Gene Details

Gene
CYP11B2
Gene Name
cytochrome P450 family 11 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000498.3 1492 Missense Mutation ATC,CTC I497L NP_000489.3

View Full Product Details