Product Details

SNP ID
rs145382016
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33500355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCCGTCCTTGAGAACAGCGGTC[C/G]AGGAGAATCAGGCAGTCTGTGGCCT
Phenotype
MIM: 614426
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MAK16 PubMed Links

Gene Details

Gene
MAK16
Gene Name
MAK16 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032509.3 1921 UTR 3 NP_115898.2
Gene
TTI2
Gene Name
TELO2 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102401.2 1921 Silent Mutation CTC,CTG L465L NP_001095871.1
NM_001265581.1 1921 Silent Mutation CTC,CTG L465L NP_001252510.1
NM_025115.3 1921 Silent Mutation CTC,CTG L465L NP_079391.2
XM_011544665.2 1921 Silent Mutation CTC,CTG L434L XP_011542967.1

View Full Product Details