Product Details
- SNP ID
-
rs145406179
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:56441292 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATATTAAAATCTCATAAATCCTC[C/T]GTATCTTTTTTCCATTTCAGGAACT
- Phenotype
-
MIM: 131330
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101929415
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34189005] are located under a probe and SNP(s) [rs3839874] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101929415
- Gene Name
- uncharacterized LOC101929415
There are no transcripts associated with this gene.
- Gene
- PENK
- Gene Name
- proenkephalin
View Full Product Details