Product Details

SNP ID
rs145490845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142703103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCCTATTAGAAATATTTCCAC[A/G]TTTTTTCATGGTTGCGAAGCAGTGC
Phenotype
MIM: 615093
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100288181 PubMed Links

Gene Details

Gene
LOC100288181
Gene Name
uncharacterized LOC100288181
There are no transcripts associated with this gene.

Gene
LY6K
Gene Name
lymphocyte antigen 6 complex, locus K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160354.1 894 UTR 3 NP_001153826.1
NM_001160355.1 894 Intron NP_001153827.1
NM_017527.3 894 Missense Mutation CAT,CGT H77R NP_059997.3
XM_005250989.4 894 Intron XP_005251046.1
XM_005250990.4 894 UTR 3 XP_005251047.1

View Full Product Details