Product Details

SNP ID
rs145610354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27241150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGCGTTGAACTCGGGAACCCC[A/C]TCAAAGGAGGGTGGCTTCAGGATGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
STMN4 PubMed Links

Gene Details

Gene
STMN4
Gene Name
stathmin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283053.1 298 Missense Mutation GAG,GAT E101D NP_001269982.1
NM_001283054.1 298 Missense Mutation GAG,GAT E74D NP_001269983.1
NM_001283055.1 298 Missense Mutation GAG,GAT E65D NP_001269984.1
NM_030795.3 298 Missense Mutation GAG,GAT E101D NP_110422.2
XM_005273652.4 298 Missense Mutation GAG,GAT E101D XP_005273709.1
XM_005273655.4 298 Missense Mutation GAG,GAT E74D XP_005273712.1

View Full Product Details