Product Details
- SNP ID
-
rs145740740
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:31644900 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCTCTTTTGATAGCTCTTAATGA[A/G]TGGGTATTTTGATTAGCTTTGTATA
- Phenotype
-
MIM: 142445
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NRG1
PubMed Links
Gene Details
- Gene
- NRG1
- Gene Name
- neuregulin 1
View Full Product Details