Product Details

SNP ID
rs145763498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:129706375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGGGAAGAAAAGGAAAAACAT[A/G]ATCTCAGAAGAAAGGAAATGCTTTA
Phenotype
MIM: 608384
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSDMC PubMed Links

Gene Details

Gene
GSDMC
Gene Name
gasdermin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031415.2 Intron NP_113603.1
XM_011517158.1 Intron XP_011515460.1
XM_011517159.1 Intron XP_011515461.1
XM_011517161.1 Intron XP_011515463.1
XM_011517162.1 Intron XP_011515464.1
XM_011517163.2 Intron XP_011515465.1
XM_011517165.1 Intron XP_011515467.1
XM_011517166.2 Intron XP_011515468.1
XM_011517167.1 Intron XP_011515469.1
XM_011517168.1 Intron XP_011515470.1
XM_017013644.1 Intron XP_016869133.1
XM_017013645.1 Intron XP_016869134.1

View Full Product Details