Product Details

SNP ID
rs145809062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:130780840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTCATACTGTGGTTTCTTAGC[A/G]CCGATAACGCCAGCTACCACTGAGC
Phenotype
MIM: 103070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY8 PubMed Links

Gene Details

Gene
ADCY8
Gene Name
adenylate cyclase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115.2 3691 Silent Mutation GGC,GGT G1102G NP_001106.1
XM_005250769.3 3691 Silent Mutation GGC,GGT G1072G XP_005250826.1
XM_006716501.3 3691 Silent Mutation GGC,GGT G1036G XP_006716564.1
XM_017013006.1 3691 Silent Mutation GGC,GGT G1006G XP_016868495.1
XM_017013007.1 3691 Intron XP_016868496.1

View Full Product Details