Product Details

SNP ID
rs145861909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123220117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATAACCCACTAAGCCAAGATTT[A/G]TATCTCTGTATGGAACTGATTTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C8orf76 PubMed Links

Gene Details

Gene
C8orf76
Gene Name
chromosome 8 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032847.2 1181 Nonsense Mutation CAA,TAA Q377* NP_116236.1
Gene
MIR4663
Gene Name
microRNA 4663
There are no transcripts associated with this gene.

Gene
ZHX1-C8orf76
Gene Name
ZHX1-C8orf76 readthrough
There are no transcripts associated with this gene.

View Full Product Details