Product Details

SNP ID
rs146156391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:43103584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCAGAACAGCAGGAGAGGCCT[C/T]GCCCCCCGAGAGGTGCCGCCAGCTG
Phenotype
MIM: 615247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POMK PubMed Links

Gene Details

Gene
POMK
Gene Name
protein-O-mannose kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277971.1 207 Silent Mutation CTC,CTT L12L NP_001264900.1
NM_032237.4 207 Silent Mutation CTC,CTT L12L NP_115613.1

View Full Product Details