Product Details

SNP ID
rs146674482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66634930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTCCAGGAGCTATTCGAATAAG[C/T]TCACTATTACTAGAAGAAGCTGTGA
Phenotype
MIM: 611745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VCPIP1 PubMed Links

Gene Details

Gene
VCPIP1
Gene Name
valosin containing protein interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025054.4 3499 Silent Mutation GAA,GAG E1080E NP_079330.2

View Full Product Details