Product Details

SNP ID
rs146889245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80632898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTAGAAACATAATAATAAATAT[C/T]ATTCATCAAAAACCCTGTTCATGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF704 PubMed Links

Gene Details

Gene
ZNF704
Gene Name
zinc finger protein 704
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033723.2 10570 UTR 3 NP_001028895.1
XM_005251279.4 10570 UTR 3 XP_005251336.2
XM_017013725.1 10570 UTR 3 XP_016869214.1

View Full Product Details