Product Details

SNP ID
rs147372571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27287666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAGGGGCGAACCTCCCCAAAGC[A/G]GGCGTGGAAGGTGTACAGGTGGCAG
Phenotype
MIM: 617007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM35 PubMed Links

Gene Details

Gene
TRIM35
Gene Name
tripartite motif containing 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304495.1 1478 UTR 3 NP_001291424.1
NM_171982.4 1478 Missense Mutation CGC,TGC R456C NP_741983.2
XM_005273452.3 1478 UTR 3 XP_005273509.1
XM_011544453.1 1478 Missense Mutation CGC,TGC R306C XP_011542755.1

View Full Product Details