Product Details
- SNP ID
-
rs147432781
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:7056647 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTTTCCTGGAGTGACTCAGCCTG[A/G]GCCTGCAGGGCCACCAGGAGAATGG
- Phenotype
-
MIM: 600472
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFA5
PubMed Links
Gene Details
- Gene
- DEFA5
- Gene Name
- defensin alpha 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021010.2 |
93 |
Silent Mutation |
GCC,GCT |
A17A |
NP_066290.1 |
View Full Product Details