Product Details

SNP ID
rs147568567
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:74986023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGCGGGAGTGGCTCAGAGTAAC[C/T]ACAGTGCTGTTCATGGCTAGAGCAA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CRISPLD1 PubMed Links

Gene Details

Gene
CRISPLD1
Gene Name
cysteine rich secretory protein LCCL domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286777.1 546 Intron NP_001273706.1
NM_001286778.1 546 Intron NP_001273707.1
NM_031461.5 546 Silent Mutation ACC,ACT T12T NP_113649.1

View Full Product Details