Product Details
- SNP ID
-
rs147945739
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143440214 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCGTGGTCCAAGGAAGCGGGAGGG[G/T]ATGAGGAGGAGGAGGAGGAGGAGGA
- Phenotype
-
MIM: 610303
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MAFA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2272753,rs2272754] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAFA
- Gene Name
- MAF bZIP transcription factor A
There are no transcripts associated with this gene.
- Gene
- ZC3H3
- Gene Name
- zinc finger CCCH-type containing 3
View Full Product Details