Product Details

SNP ID
rs148003710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11996223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCACTGGATCCCAAAGGAAAAGA[A/G]AGACAAACACAACAAGGAGAGGCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEFB134 PubMed Links

Gene Details

Gene
DEFB134
Gene Name
defensin beta 134
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302695.1 121 Missense Mutation TCT,TTT S10F NP_001289624.1
XM_017013724.1 121 Missense Mutation TCT,TTT S10F XP_016869213.1

View Full Product Details