Product Details

SNP ID
rs148058456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6710287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTTCTGAAATTGCATACCTTA[C/T]CTGATGTTCAGAGATCCGATTTACT
Phenotype
MIM: 614796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGPAT5 PubMed Links

Gene Details

Gene
AGPAT5
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018361.3 Intron NP_060831.2
XM_011534751.2 Intron XP_011533053.1
Gene
MCPH1-AS1
Gene Name
MCPH1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details