Product Details

SNP ID
rs148153750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:116649894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGAAAGTGAAAGATGAGTCAGC[C/T]ATGCCAAAAATATTGTAGGCAAAAG
Phenotype
MIM: 603912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3H PubMed Links

Gene Details

Gene
EIF3H
Gene Name
eukaryotic translation initiation factor 3 subunit H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003756.2 Intron NP_003747.1
Gene
LOC105375713
Gene Name
uncharacterized LOC105375713
There are no transcripts associated with this gene.

View Full Product Details