Product Details

SNP ID
rs148299130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144475218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGAGTCTTTCCAGCCCTCGTAG[G/T]CTTCCCTGAAGAAGGGGAACACGGC
Phenotype
MIM: 616635 MIM: 603621 MIM: 615216 MIM: 609172
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYHR1 PubMed Links

Gene Details

Gene
CYHR1
Gene Name
cysteine and histidine rich 1
There are no transcripts associated with this gene.

Gene
FOXH1
Gene Name
forkhead box H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003923.2 797 Missense Mutation GAC,GCC D73A NP_003914.1
Gene
KIFC2
Gene Name
kinesin family member C2
There are no transcripts associated with this gene.

Gene
PPP1R16A
Gene Name
protein phosphatase 1 regulatory subunit 16A
There are no transcripts associated with this gene.

View Full Product Details