Product Details

SNP ID
rs148496185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:107251969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCATGGTTTTGTCCCGCAGTATA[A/G]AACATTCCATTTAGATTGGAGGGGC
Phenotype
MIM: 601667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANGPT1 PubMed Links

Gene Details

Gene
ANGPT1
Gene Name
angiopoietin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146.4 1062 Silent Mutation NP_001137.2
NM_001199859.2 1062 Silent Mutation NP_001186788.1
NM_001314051.1 1062 Silent Mutation NP_001300980.1

View Full Product Details