Product Details

SNP ID
rs148537365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27287619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCAAAGGCTCTGGAGGCCCGGC[A/G]CCCCGTGCACCCCCCAGGTAGAAGT
Phenotype
MIM: 617007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM35 PubMed Links

Gene Details

Gene
TRIM35
Gene Name
tripartite motif containing 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304495.1 1525 UTR 3 NP_001291424.1
NM_171982.4 1525 Silent Mutation GGC,GGT G471G NP_741983.2
XM_005273452.3 1525 UTR 3 XP_005273509.1
XM_011544453.1 1525 Silent Mutation GGC,GGT G321G XP_011542755.1

View Full Product Details