Product Details

SNP ID
rs148577927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6870722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCCCTCTGTAACAGGTGCCTT[A/G]AATTTTGGTAAAGATCGGGCAGGCA
Phenotype
MIM: 602056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEFB1 PubMed Links

Gene Details

Gene
DEFB1
Gene Name
defensin beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005218.3 316 Nonsense Mutation CAA,TAA Q56* NP_005209.1

View Full Product Details