Product Details

SNP ID
rs148618129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93732387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGAAGTCAAAATTCAACCACA[C/G]AAGTCTTAACAGTTATAGAGTCTTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM92A1 PubMed Links

Gene Details

Gene
FAM92A1
Gene Name
family with sequence similarity 92 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283034.1 4217 Intron NP_001269963.1
NM_145269.4 4217 Intron NP_660312.2
XM_005250787.2 4217 Intron XP_005250844.1
Gene
RBM12B
Gene Name
RNA binding motif protein 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203390.2 4217 UTR 3 NP_976324.2
XM_005250915.4 4217 UTR 3 XP_005250972.1
XM_011517027.2 4217 UTR 3 XP_011515329.1
XM_011517028.2 4217 UTR 3 XP_011515330.1
XM_011517029.2 4217 UTR 3 XP_011515331.1
XM_017013406.1 4217 UTR 3 XP_016868895.1
XM_017013407.1 4217 UTR 3 XP_016868896.1
XM_017013408.1 4217 UTR 3 XP_016868897.1
Gene
RBM12B-AS1
Gene Name
RBM12B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details