Product Details

SNP ID
rs149544104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124311337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTAGTAACAGAATATACCAAG[A/G]CCCTCTGAGATTAGTAAGAAATCAA
Phenotype
MIM: 616609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM65 PubMed Links

Gene Details

Gene
TMEM65
Gene Name
transmembrane protein 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194291.2 3888 UTR 3 NP_919267.2
XM_011516847.2 3888 Intron XP_011515149.1
XM_017013104.1 3888 Intron XP_016868593.1
XM_017013105.1 3888 Intron XP_016868594.1

View Full Product Details