Product Details
- SNP ID
-
rs149678009
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:124450979 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGGAGAGAGAAAGTGGGAAGCCC[A/G]TGGCTGTTGTCGCAGTTGTGACTGA
- Phenotype
-
MIM: 611244
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRMT12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3812475] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRMT12
- Gene Name
- tRNA methyltransferase 12 homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017956.3 |
173 |
Missense Mutation |
ATG,GTG |
M18V |
NP_060426.2 |
View Full Product Details