Product Details

SNP ID
rs149835506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:145052811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACAAAAGAATAAGAAGAAAACG[C/T]GGAACAGGGCCTCTGTGGCAAATGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf33 PubMed Links

Gene Details

Gene
C8orf33
Gene Name
chromosome 8 open reading frame 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023080.2 286 Missense Mutation CGG,TGG R78W NP_075568.1
Gene
LOC107983994
Gene Name
endogenous retrovirus group K member 9 Pol protein-like
There are no transcripts associated with this gene.

View Full Product Details