Product Details

SNP ID
rs150222936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144040018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCTAACCAGCTTGCAGCCCAG[C/T]GCCACACCGGGCTCACTCATGAGCC
Phenotype
MIM: 610874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMPD5 PubMed Links

Gene Details

Gene
SMPD5
Gene Name
sphingomyelin phosphodiesterase 5
There are no transcripts associated with this gene.

Gene
SPATC1
Gene Name
spermatogenesis and centriole associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134374.1 423 Silent Mutation AGC,AGT S107S NP_001127846.1
NM_198572.2 423 Silent Mutation AGC,AGT S107S NP_940974.2
XM_011517021.1 423 Silent Mutation AGC,AGT S107S XP_011515323.1
XM_011517022.2 423 Silent Mutation AGC,AGT S101S XP_011515324.1
XM_011517023.2 423 Silent Mutation AGC,AGT S45S XP_011515325.1
XM_011517024.2 423 Silent Mutation AGC,AGT S16S XP_011515326.1
XM_011517025.2 423 Silent Mutation AGC,AGT S16S XP_011515327.1
XM_017013399.1 423 Silent Mutation AGC,AGT S16S XP_016868888.1

View Full Product Details