Product Details

SNP ID
rs150264603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142750346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGCAGTGGAGGCTGTTCAGAGC[A/G]GGCGCCCCGTCTACATTGCACAGCT
Phenotype
MIM: 606119
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYPD2 PubMed Links

Gene Details

Gene
LYPD2
Gene Name
LY6/PLAUR domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205545.2 398 Silent Mutation CCC,CCT P105P NP_991108.1
Gene
SLURP1
Gene Name
secreted LY6/PLAUR domain containing 1
There are no transcripts associated with this gene.

View Full Product Details