Product Details

SNP ID
rs150274198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:74986088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCAATGCCACTTTATTGGAGA[A/G]ACTTTTGGAAAAATACATGGATGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRISPLD1 PubMed Links

Gene Details

Gene
CRISPLD1
Gene Name
cysteine rich secretory protein LCCL domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286777.1 611 Intron NP_001273706.1
NM_001286778.1 611 Intron NP_001273707.1
NM_031461.5 611 Missense Mutation AAA,AGA K34R NP_113649.1

View Full Product Details