Product Details

SNP ID
rs150282653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42838107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGCCTTCTGCATCGCTGCTGT[A/G]CGGTCTTGAGCTTCTTTCTGAGTTT
Phenotype
MIM: 609520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THAP1 PubMed Links

Gene Details

Gene
THAP1
Gene Name
THAP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018105.2 734 Missense Mutation GCA,GTA A166V NP_060575.1
NM_199003.1 734 UTR 3 NP_945354.1

View Full Product Details