Product Details

SNP ID
rs150378190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:99891960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCACGAATGCTACAGCCATATGA[C/T]TTCGCAGACGCCTGGCCAGAAGGCC
Phenotype
MIM: 124090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX6C PubMed Links

Gene Details

Gene
COX6C
Gene Name
cytochrome c oxidase subunit 6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004374.3 469 Missense Mutation AAT,AGT N21S NP_004365.1
XM_017013020.1 469 Intron XP_016868509.1

View Full Product Details