Product Details

SNP ID
rs150400388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22605806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTAAGCGCCAGCGGATCAACCC[A/G]CTTCCAGGGGGACGCAACTTCTCAG
Phenotype
MIM: 607359 MIM: 609722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C8orf58 PubMed Links

Gene Details

Gene
C8orf58
Gene Name
chromosome 8 open reading frame 58
There are no transcripts associated with this gene.

Gene
CCAR2
Gene Name
cell cycle and apoptosis regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021174.5 282 Silent Mutation CCA,CCG P11P NP_066997.3
XM_011544603.1 282 Silent Mutation CCA,CCG P11P XP_011542905.1
XM_011544604.1 282 Silent Mutation CCA,CCG P11P XP_011542906.1
XM_017013717.1 282 Silent Mutation CCA,CCG P11P XP_016869206.1
Gene
LOC107986876
Gene Name
proline-rich proteoglycan 2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017014122.1 282 Intron XP_016869611.1
Gene
PDLIM2
Gene Name
PDZ and LIM domain 2
There are no transcripts associated with this gene.

View Full Product Details