Product Details

SNP ID
rs150892020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22690518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGCCAGCGACACGGGGGGCGC[C/T]GAGGATGCAGAGGGTGCACCGCCCT
Phenotype
MIM: 602419
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EGR3 PubMed Links

Gene Details

Gene
EGR3
Gene Name
early growth response 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199880.1 1428 Silent Mutation NP_001186809.1
NM_001199881.1 1428 Silent Mutation NP_001186810.1
NM_004430.2 1428 Silent Mutation NP_004421.2
XM_005273425.3 1428 Silent Mutation XP_005273482.1
XM_005273426.3 1428 Silent Mutation XP_005273483.1
XM_011544429.2 1428 Silent Mutation XP_011542731.1

View Full Product Details