Product Details

SNP ID
rs150929068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:102551922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATTCCAAGCGATCACGCTCTTG[C/T]GGCCTGTGTGATCTCTACCCATGTT
Phenotype
MIM: 182878
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ODF1 PubMed Links

Gene Details

Gene
ODF1
Gene Name
outer dense fiber of sperm tails 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024410.3 303 Silent Mutation TGC,TGT C65C NP_077721.2

View Full Product Details