Product Details

SNP ID
rs150942595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81283457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAAAACCTCACCATAAAAACTGA[A/G]AGCACTTTGAAAACAACACAGTTTT
Phenotype
MIM: 605168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FABP5 PubMed Links

Gene Details

Gene
FABP5
Gene Name
fatty acid binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001444.2 Intron NP_001435.1

View Full Product Details