Product Details

SNP ID
rs151129845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39746584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAAGGAATGAATAAGAAAAATG[C/G]CCATCTCATTGGTTTGGAATGGTAA
Phenotype
MIM: 601533
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAM2 PubMed Links

Gene Details

Gene
ADAM2
Gene Name
ADAM metallopeptidase domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278113.1 2014 Missense Mutation CCA,GCA P669A NP_001265042.1
NM_001278114.1 2014 Missense Mutation CCA,GCA P625A NP_001265043.1
NM_001464.4 2014 Missense Mutation CCA,GCA P688A NP_001455.3
XM_005273468.1 2014 Missense Mutation CCA,GCA P658A XP_005273525.1
XM_011544479.1 2014 Missense Mutation CCA,GCA P562A XP_011542781.1
XM_011544480.1 2014 Missense Mutation CCA,GCA P532A XP_011542782.1

View Full Product Details