Product Details
- SNP ID
-
rs151255540
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:10898246 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCCTGTTGTTCCGTTACGGCTCT[C/G]GTGGGCGTAACCTGGGTCCCCCGGT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
XKR6
PubMed Links
Gene Details
- Gene
- XKR6
- Gene Name
- XK related 6
View Full Product Details