Product Details

SNP ID
rs151302692
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26505500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACAGGTCTGATCTACACAACCA[C/T]GTGGTGCTACCGCTTTTAAAATCTG
Phenotype
MIM: 602463 MIM: 603970
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DPYSL2 PubMed Links

Gene Details

Gene
DPYSL2
Gene Name
dihydropyrimidinase like 2
There are no transcripts associated with this gene.

Gene
PNMA2
Gene Name
paraneoplastic Ma antigen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007257.5 4026 UTR 3 NP_009188.1
XM_011544365.2 4026 UTR 3 XP_011542667.1

View Full Product Details