Product Details

SNP ID
rs137939984
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94603524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCAGTGGTGGCCATTCCCCCAG[A/C]CTTCTCCATGACGTAGGCCATGGGG
Phenotype
MIM: 611570 MIM: 603027
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
FBP1 PubMed Links

Gene Details

Gene
FBP1
Gene Name
fructose-bisphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000507.3 1205 Missense Mutation GCT,TCT A292S NP_000498.2
NM_001127628.1 1205 Missense Mutation GCT,TCT A292S NP_001121100.1
XM_006717005.3 1205 Missense Mutation GCT,TCT A210S XP_006717068.1
Gene
FBP2
Gene Name
fructose-bisphosphatase 2
There are no transcripts associated with this gene.

View Full Product Details