Product Details

SNP ID
rs138140692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33922584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCAGGGAGCTGGGCTGGCTGC[A/G]CTGACCCGAGCCACTCTGAGGAAGA
Phenotype
MIM: 612506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBAP2 PubMed Links

Gene Details

Gene
UBAP2
Gene Name
ubiquitin associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282529.1 3393 Missense Mutation CGC,TGC R827C NP_001269458.1
NM_001282530.1 3393 Missense Mutation CGC,TGC R333C NP_001269459.1
NM_018449.3 3393 Missense Mutation CGC,TGC R1094C NP_060919.3
Gene
UBE2R2
Gene Name
ubiquitin conjugating enzyme E2 R2
There are no transcripts associated with this gene.

View Full Product Details