Product Details

SNP ID
rs138570823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122750755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAAGATATGAAGAGGGGTTTGA[A/C]GAAATTACAGGACAGAATTTACCGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR1L6 PubMed Links

Gene Details

Gene
OR1L6
Gene Name
olfactory receptor family 1 subfamily L member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004453.2 908 Missense Mutation AAG,ACG K303T NP_001004453.2

View Full Product Details